Learn More
Cayman Chemical RCS4 N 5carboxypentyl met 1mg
SDP
Catalog No. 502951177
Encompass_Preferred
An expected metabolite of RCS-4, which should be detectable in the urine
Catalog No. 50-295-1177 Supplier Cayman Chemical Supplier No. 109371 MG
An expected metabolite of RCS-4, which should be detectable in the urine