Learn More
Cayman Chemical RCS4 N 5hydroxypentyl met 1mg
Supplier: Cayman Chemical 143641 MG
An internal standard for the quantification of RCS-4 N-(5-hydroxypentyl) metabolite by GC- or LC-MS; intended for research and forensic applications.