Learn More
Cayman Chemical RCS4 NpentnoIc ACD metabo
SDP
Catalog No. 502953938
Encompass_Preferred
An internal standard for the quantification of RCS-4 N-pentanoic acid metabolite by GC- or LC-MS; intended for research and forensic applications
Catalog No. 50-295-3938 Supplier Cayman Chemical Supplier No. 14365100 UG
An internal standard for the quantification of RCS-4 N-pentanoic acid metabolite by GC- or LC-MS; intended for research and forensic applications