Learn More
Cayman Chemical RCS4C4 homolog Analytical Refe
SDP
Supplier:  Cayman Chemical 107985 MG
Encompass_Preferred
Identical to RCS-4 except the N-1 alkyl chain length has been shortened from C5 to C4; detected in herbal blends
Catalog No. 50-288-7385