Learn More
Cayman Chemical RCS-4-C4 homolog, Purity: ≥98%, CAS: 1345966-77-9, 1 mg
SDP
Catalog No. 502888820
Encompass_Preferred
Identical to RCS-4 except the N-1 alkyl chain length has been shortened from C5 to C4; detected in herbal blends
Catalog No. 50-288-8820 Supplier Cayman Chemical Supplier No. 107981 MG
Identical to RCS-4 except the N-1 alkyl chain length has been shortened from C5 to C4; detected in herbal blends