Learn More
JASCO INC RCT QUARTZ CELL W/STOPPER 2MM
SDP
Catalog No. NC2124192
Encompass
Rectangular Quartz Cell-2mm pathlengthwith PTFE stopper
This item is not returnable. View return policy
Catalog No. NC2124192 Supplier JASCO INC Supplier No. 11034152
Rectangular Quartz Cell-2mm pathlengthwith PTFE stopper