Learn More
Electron Microscopy Sciences Gilder Rectangular Slotted Grid G100/400, Nickel
SDP
Catalog No. 5028323
Encompass_Preferred
A reliable support specimen grid source. Features: well-defined grid bars, maximum open area, and a matt/shiny side. Each grid is individually inspected. 3.05 mm diameter, 0.7 mil (18um) thickness.
Material: Nickel
Pitch: 250/62 µm
Hole: 205/37 µm
Bar: 45/25 µm
Catalog No. 50-283-23 Supplier Electron Microscopy Sciences Supplier No. G1040NI
A reliable support specimen grid source. Features: well-defined grid bars, maximum open area, and a matt/shiny side. Each grid is individually inspected. 3.05 mm diameter, 0.7 mil (18um) thickness.
Material: Nickel
Pitch: 250/62 µm
Hole: 205/37 µm
Bar: 45/25 µm