Learn More
Electron Microscopy Sciences Veco Grid L0.2 x 0.5, Nickel with 200 x 500 µm Rectangle Slot
SDP
Catalog No. 50980129
Encompass_Preferred
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter.
Material: Nickel
Slot: 200 x 500 µm
Catalog No. 50-980-129 Supplier Electron Microscopy Sciences Supplier No. 0502NI
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter.
Material: Nickel
Slot: 200 x 500 µm