Learn More
Electron Microscopy Sciences Gilder Rectangular Slotted Grid G75/300, Copper
SDP
Catalog No. 5028321
Encompass_Preferred
A reliable support specimen grid source. Features: well-defined grid bars, maximum open area, and a matt/shiny side. Each grid is individually inspected. 3.05 mm diameter, 0.7 mil (18um) thickness.
Material: Copper
Pitch: 340/83 µm
Hole: 290/58 µm
Bar: 50/25 µm
Catalog No. 50-283-21 Supplier Electron Microscopy Sciences Supplier No. G7530CU
A reliable support specimen grid source. Features: well-defined grid bars, maximum open area, and a matt/shiny side. Each grid is individually inspected. 3.05 mm diameter, 0.7 mil (18um) thickness.
Material: Copper
Pitch: 340/83 µm
Hole: 290/58 µm
Bar: 50/25 µm