Learn More
Electron Microscopy Sciences Veco Grid L2 x 0.6, Nickel with 2000 x 600 µm Rectangle Slot
SDP
Catalog No. 50980133
Encompass_Preferred
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter.
Material: Nickel
Slot: 2000 x 600 µm
Catalog No. 50-980-133 Supplier Electron Microscopy Sciences Supplier No. 0620NI
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter.
Material: Nickel
Slot: 2000 x 600 µm