Learn More
Aviva Systems Biology Corp RCV1 Antibody - C-terminal region (ARP38196_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38196_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RCV1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human RCV1. Alternate names: RCV1, RCVRNApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Pig, Rabbit, Rat
Catalog No. 50-014-56558