Learn More
Chemglass Life Sciences Cover Slip, Round, 25mm Dia, German Glass, #1 Thickness, 1oz Pkg, Approx 160/Oz
SDP
Catalog No. 501215161
Encompass_Preferred
Cover Slip, Round, 25mm Dia, German Glass, #1 Thickness, 1oz Pkg. Cover slips are manufactured from the highest quality borosilicate hydrolytic class 1 German glass. They have excellent resistance to chemicals and are ideal for neural tissue culture and hard-to-attach cells. These cover slips are exceptional for florescence and confocal microscopy. Sold in packages of 1oz.Please Note:25mm Diameter will fit in a 6 well plate; CLS-3500-006Approximate Quantities per Case:CLS-1760-025: 160
Catalog No. 50-121-5161 Supplier Chemglass Life Sciences Supplier No. CLS1760025
Cover Slip, Round, 25mm Dia, German Glass, #1 Thickness, 1oz Pkg. Cover slips are manufactured from the highest quality borosilicate hydrolytic class 1 German glass. They have excellent resistance to chemicals and are ideal for neural tissue culture and hard-to-attach cells. These cover slips are exceptional for florescence and confocal microscopy. Sold in packages of 1oz.Please Note:25mm Diameter will fit in a 6 well plate; CLS-3500-006Approximate Quantities per Case:CLS-1760-025: 160