Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RD Plastics Clear Drawstring Bag
SDP

Catalog No. 23700227
Change view
Click to view available options
Dimensions (L x W):
18 x 15 in.
23 x 17.5 in.
Product Type:
Biohazard Bag
Drawstring Bag
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W) Product Type
23-700-227 18 x 15 in. Drawstring Bag
23-700-235 23 x 17.5 in. Biohazard Bag
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23-700-227 Supplier RD Plastics Supplier No. G127
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Clear drawstring bags allow for easy content identification and keep contents secure. Available unprinted or with histology biohazard markings.

  • Clear LDPE transport bags with nylon drawstring closure
  • Allows for easy content identification and holds contents securely
  • Unmarked or printed with “HISTOLOGY TISSUE” and orange and black biohazard symbol

Specifications

Autoclavable Not Autoclavable
Closure Type Drawstring
Color Clear
For Use With (Application) For easy content identification and keeps contents secure
Material LDPE
No. per Case 500/Cs.
Thickness (English) 2 mil
Dimensions (L x W) 18 x 15 in.
Product Type Drawstring Bag
Capacity (English) 7 lb.
Length (Metric) 45.72 cm
Width (Metric) 38.1 cm
Thickness (Metric) 0.051 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.