Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RD Plastics Absorbent Pad

Catalog No. 23675320
Change view
Click to view available options
Length (English):
3 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
23-675-320 3 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-675-320 Supplier RD Plastics Supplier No. L16
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Impregnated with super-absorbent polymers, it is for use with specimen transport bags

  • Poly-coated backing for protection against fluid penetration
  • 7.62 × 15.24cm (3 × 6 in.) dimensions

Specifications

Material Polymer
Type Absorbent Pad
For Use With (Application) Specimen transport bags
Length (English) 3 in.
Length (Metric) 7.62 cm
Width (English) 6 in.
Width (Metric) 15.24 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.