Learn More
RD Plastics LABTITE™ Transport Bags
Catalog No. 23675321
Change view
Click to view available options
Length (English):
10 in.
Catalog No. Length (English)
23-675-321 10 in.
1 options
Catalog No. 23-675-321 Supplier RD Plastics Supplier No. G119
Add to Cart
Add to Cart
Liquid-tight adhesive closure system
  • 3-wall construction
  • Printed with biohazard symbol
  • Tamper evident tape closure with pull-away blue release line
  • Perforated for ease of separation
Order Info
Shipped 1000 per case
Specifications
Description Orange and black biohazard symbol and storage instructions
Closure Type Tamper-evident Tape
Color Clear
Includes Three wall construction, Perforated bottom
No. per Case 1000/Cs.
Length (English) 10 in.
Width (English) 10 in.
Product Type Biohazard Specimen Transport Bag