Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RD Plastics LABTITE™ Tranport Bag

Catalog No. 23700241
Change view
Click to view available options
Dimensions (L x W):
10 x 6 in.
18 x 13 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
23-700-241 18 x 13 in.
23-700-206 10 x 6 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23-700-241 Supplier RD Plastics Supplier No. G117
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

It features a liquid-tight adhesive closure system

  • Pull away release liner
  • Press to close liner
  • No glove-to adhesive contact
  • Three wall construction
  • Liquid-tight closure
  • Designed polymer film

Specifications

Color Clear
For Use With (Application) Used for specimen transport
Includes Printed with biohazard symbol, closure pull away blue release line and seal bag
Thickness (English) 2 mil
Dimensions (L x W) 18 x 13 in.
Length (English) 18 in.
Product Type Biohazard Specimen Transport Bag
Length (Metric) 45.7 cm
Thickness (Metric) 0.279 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.