Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RD Plastics Reclosable Specimen Transport Bag

Catalog No. 23700213
Change view
Click to view available options
Dimensions (L x W):
6 x 4 in.
6 x 9 in.
9 x 6 in.
10 x 8 in.
10 x 10 in.
Includes:
2-wall Construction, Storage Instructions
3-wall Construction, Storage Instructions
Absorbent Pad
Absorbent Pad, STAT Printed
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W) Includes
23-700-213 9 x 6 in. Absorbent Pad, STAT Printed
23-700-212 9 x 6 in. 3-wall Construction, Storage Instructions
23-700-207 6 x 9 in. 2-wall Construction, Storage Instructions
23-700-236 6 x 4 in. 2-wall Construction, Storage Instructions
23-700-208 10 x 8 in. Absorbent Pad
01-800-102 10 x 10 in. 2-wall Construction, Storage Instructions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 23-700-213 Supplier RD Plastics Supplier No. Q793
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Light duty and available in both two or three wall construction

  • Specimen transport
  • Two wall or three wall
  • Flap cover to protect papers
  • Biohazard print
  • Score line for easy opening
  • Absobent pad

Specifications

Closure Type Reclosable
For Use With (Application) For specimen transport
Includes Absorbent Pad, STAT Printed
Thickness (English) 2 mil
Dimensions (L x W) 9 x 6 in.
Length (English) 9 in.
Width (English) 6 in.
Product Type Reclosable Specimen Transport Bag
Length (Metric) 22.8 cm
Width (Metric) 15.2 cm
Thickness (Metric) 0.279 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.