Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RD Plastics Biohazard Ziplock Bag, Green Tint

Catalog No. 23700242
Change view
Click to view available options
Dimensions (L x W):
15 x 12 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
23-700-242 15 x 12 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-700-242 Supplier RD Plastics Supplier No. Q393
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Color coded biohazard bag for easy identification of the contents.

  • LDPE bag in 2 wall style with 2 mil thickness
  • Green tinted bag printed with orange and black biohazard symbol
  • Features a resealable zipper closure

Specifications

Autoclavable Not Autoclavable
Color Green
Material LDPE
No. per Case 1000
No. per Pack 100
Thickness (English) 2 mil
Dimensions (L x W) 15 x 12 in.
Capacity (English) 4 lb.
Length (Metric) 38.1 cm
Width (Metric) 30.48 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.