Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RD Plastics Specimen Transport Bag

Catalog No. 21465300
Change view
Click to view available options
Dimensions (L x W):
12 x 12.75 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
21-465-300 12 x 12.75 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-465-300 Supplier RD Plastics Supplier No. B38
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Die cut handle biohazard ziplock specimen transport bag with biohazard print.

Features a gusseted bottom to keep it in an upright standing position.

Specifications

Autoclavable Not Autoclavable
Color Assorted
Material LDPE
No. per Case 500
Dimensions (L x W) 12 x 12.75 in.
Capacity (English) 5 lb.
Length (Metric) 30.48 cm
Width (Metric) 32.38 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.