Learn More
RD Plastics Three-wall Reclosable Biohazard Specimen Transport Bags
Description
- Three-wall construction
- Pouch has a flap cover to hold paperwork stays in place
- With orange and black biohazard symbol and storage instructions
Specifications
Specifications
| Closure Type | Reclosable |
| Color | Clear |
| For Use With (Application) | Used for specimen transport |
| Includes | Pouch with flap cover, Biohazard symbol and storage instructions |
| Thickness (English) | 2 mil |
| Dimensions (L x W) | 9 x 6 in. |
| Length (English) | 9 in. |
| Width (English) | 6 in. |
| Product Type | Biohazard Specimen Transport Bag (Light Duty) |
| Length (Metric) | 22.86 cm |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.