Learn More
MSC Markal Welder's Pencil, Metallic Silver, Round Tip, 3/Pk.
Catalog No. 501113127
Encompass
This welder's pencil is designed for marking on various metals in welding environments. It provides clear, easy-to-see silver marks that are more durable than chalk or soapstone and suitable for layout and fabrication tasks.
  • Leaves an easy-to-see silver mark
  • Does not wipe off as easily as chalk or soapstone
  • Can be removed with a wire brush or most solvents/cleaners
  • Shows up well in low light conditions
  • Suitable for steel, iron, aluminum, and stainless steel
  • Ideal for layout work, welding prep, and joint and hole location marking
  • Supplied as a 3 piece kit
Catalog No. 50-111-3127 Supplier MSC Supplier No. 60244332
This welder's pencil is designed for marking on various metals in welding environments. It provides clear, easy-to-see silver marks that are more durable than chalk or soapstone and suitable for layout and fabrication tasks.
  • Leaves an easy-to-see silver mark
  • Does not wipe off as easily as chalk or soapstone
  • Can be removed with a wire brush or most solvents/cleaners
  • Shows up well in low light conditions
  • Suitable for steel, iron, aluminum, and stainless steel
  • Ideal for layout work, welding prep, and joint and hole location marking
  • Supplied as a 3 piece kit