Learn More
Aviva Systems Biology Corp RD3 Antibody - N-terminal region (ARP55856_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55856_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RD3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human RD3. Alternate names: C1orf36, LCA12ApplicationWB. Species reactivity: Dog, Human
Catalog No. 50-014-61637