Learn More
Aviva Systems Biology Corp RDBP Antibody - N-terminal region (ARP40431_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40431_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RDBP. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human RDBP. Alternate names: D6S45, NELF-E, RD, RDP, RDBPApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast
Catalog No. 50-014-57448