Learn More
ELEMENTAR AMERICAS INC RDCTN TB QRTZ LBLD WTH FIL
SDP
Catalog No. NC2736095
Encompass
  • Type: Reduction tube
  • Material: Quartz
  • Feature: Labeled
  • Accessory: Filling instructions included
  • Application: CHNS/CHN analysis
  • Compatibility: UNICUBE, vario MICRO cube, vario EL cube, vario ISOTOPE select, vario ISOTOPE cube
Catalog No. NC2736095 Supplier ELEMENTAR AMERICAS INC Supplier No. 100005770
  • Type: Reduction tube
  • Material: Quartz
  • Feature: Labeled
  • Accessory: Filling instructions included
  • Application: CHNS/CHN analysis
  • Compatibility: UNICUBE, vario MICRO cube, vario EL cube, vario ISOTOPE select, vario ISOTOPE cube