Learn More
Key Scientific Products RDE II Receptor Destroying Enzyme, Eliminates Nonspecific Hemagglutination Inhibitors, IgE Activity Modulation, 5 vials x 20 mL
SDP
Supplier:  Key Scientific Products M370013
Encompass_Preferred
RDE II Receptor Destroying Enzyme is an innovative lyophilized enzyme designed to eliminate nonspecific hemagglutination inhibitors in human serum, enhancing the accuracy of hemagglutination inhibition (HI) testing for influenza viruses. By preventing false positives due to serum factors, it ensures reliable results. Additionally, it can modulate IgE activity, opening new avenues for allergy research. Derived from *Vibrio cholerae*, it stands out in virology applications, intended for research use only.
Catalog No. NC1157652
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
View Alternate Product