Learn More
MOLDPRO INC Sample Holder for RDE-OES, ~2 mL, 18 x 9.5 mm, Black, 1000/pkg, Flat bottom, PP Homopolymer
Catalog No. NC2233821
Encompass_Preferred
MP-116 Sample holder is made to fit SpectrOil M Series Rotating Disc Electrode Optical Emission Spectroscope (RDEOES). Comparable to Spectro Scientific M90200 & P-10524, SPC Science 070-010-101 and NSN 6640-01-445-1330. Made with Polypropylene homopolymer and packaged 1000 cups/pack.• Used for sample presentation to SpectrOil M/N-W, M/C-W, M/F-W, M/R and 100 series instruments• Can be used manually or with Double Disc Rapid Robot (D2R2)• Used for ASTM D6595 and ASTM D6728 test methods
This item is not returnable. View return policy
Catalog No. NC2233821 Supplier MOLDPRO INC Supplier No. MP116
MP-116 Sample holder is made to fit SpectrOil M Series Rotating Disc Electrode Optical Emission Spectroscope (RDEOES). Comparable to Spectro Scientific M90200 & P-10524, SPC Science 070-010-101 and NSN 6640-01-445-1330. Made with Polypropylene homopolymer and packaged 1000 cups/pack.• Used for sample presentation to SpectrOil M/N-W, M/C-W, M/F-W, M/R and 100 series instruments• Can be used manually or with Double Disc Rapid Robot (D2R2)• Used for ASTM D6595 and ASTM D6728 test methods