Learn More
Aviva Systems Biology Corp RDH10 Antibody - N-terminal region (ARP53216_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53216_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RDH10. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human RDH10. Alternate names: SDR16C4ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60976