Learn More
Aviva Systems Biology Corp RDH11 Antibody - C-terminal region (ARP48350_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48350_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RDH11. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human RDH11. Alternate names: ARSDR1, CGI-82, FLJ32633, HCBP12, MDT1, PSDR1, RALR1, SCALD, CGI82, SDR7C1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-59801