Learn More
Aviva Systems Biology Corp RDH13 Antibody - C-terminal region (ARP62403_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62403_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RDH13. It was validated on Western Blot. . Alternate names: DKFZp313H0740, FLJ39479, SDR7C3ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-287-673