Learn More
Aviva Systems Biology Corp RDH16 Antibody - middle region (ARP42142_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42142_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RDH16. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human RDH16. Alternate names: RODH-4, SDR9C8ApplicationWB. Species reactivity: Dog, Horse, Human, Rabbit
Catalog No. 50-284-978