Learn More
Grainger Mr. Heater MH4B Portable Radiant Propane Heater, 3,800 BtuH, 1 ea
Catalog No. 502786414
Encompass_Preferred
This patented radiant 3,800 BTU liquid propane heater is designed for heating small enclosed spaces up to 95 sq. ft. It connects directly to a 1 lb. disposable propane cylinder and features an integrated handle for portability. The unit utilizes a matchless piezo ignition system for simple, one-button operation, providing quiet and efficient heating performance.
  • 3,800 BTU heat output
  • Oxygen depletion sensor (ODS) and accidental tip-over safety shut-off
  • Matches piezo ignition system
  • Connects directly to 1 lb. disposable propane cylinder
  • Includes a space-saving round stand
  • Provides up to 5.6 hours of run time
  • Certified for safe use in accordance with ANSI and CSA standards
Catalog No. 50-278-6414 Supplier Grainger Supplier No. 197JM4
This patented radiant 3,800 BTU liquid propane heater is designed for heating small enclosed spaces up to 95 sq. ft. It connects directly to a 1 lb. disposable propane cylinder and features an integrated handle for portability. The unit utilizes a matchless piezo ignition system for simple, one-button operation, providing quiet and efficient heating performance.
  • 3,800 BTU heat output
  • Oxygen depletion sensor (ODS) and accidental tip-over safety shut-off
  • Matches piezo ignition system
  • Connects directly to 1 lb. disposable propane cylinder
  • Includes a space-saving round stand
  • Provides up to 5.6 hours of run time
  • Certified for safe use in accordance with ANSI and CSA standards