Learn More
Grainger Mr. Heater Portable Radiant Propane Heater, 12,000 BtuH, 1 ea
Catalog No. 502786415
Encompass_Preferred
This portable radiant propane heater is engineered to deliver efficient, clean heat in a compact design. It features a versatile fuel system that connects directly to a 1 lb. propane cylinder or to larger fuel sources via a hose, making it suitable for heating areas up to 300 sq. ft. Built with safety and convenience in mind, the unit is equipped with integrated security sensors and matchless ignition.
  • Oxygen depletion sensor and accidental tip-over safety shut-off systems
  • Silent electronic piezo ignition for reliable, quiet startup
  • Swivel regulator allows for use with various tank sizes
  • Portable design ideal for camping, ice fishing, and remote work applications
  • Variable output settings from 6,000 to 12,000 BtuH
Catalog No. 50-278-6415 Supplier Grainger Supplier No. 197JN0
This portable radiant propane heater is engineered to deliver efficient, clean heat in a compact design. It features a versatile fuel system that connects directly to a 1 lb. propane cylinder or to larger fuel sources via a hose, making it suitable for heating areas up to 300 sq. ft. Built with safety and convenience in mind, the unit is equipped with integrated security sensors and matchless ignition.
  • Oxygen depletion sensor and accidental tip-over safety shut-off systems
  • Silent electronic piezo ignition for reliable, quiet startup
  • Swivel regulator allows for use with various tank sizes
  • Portable design ideal for camping, ice fishing, and remote work applications
  • Variable output settings from 6,000 to 12,000 BtuH