Learn More
Electron Microscopy Sciences RDO Specimen Decalcifier 1 LT
SDP
Catalog No. 5033454
Encompass_Preferred
RDO Decalcifier was developed with the basic premise of making your work routine quicker and easier without sacraficing the quality and integrity of your samples. It is a remarkably effective multipurpose decacifier that can be tailored to suit your specific lab routine. RDO is safe, non-toxic, and non-corrisive. It is dilutable, microwavable, biodegradable, and translucent.
RDO Decalcifier offers crisp nuclear staining and definition, gives distinguishable chromatin and shows great surface decalcification. It is safe enough to be disposed down lab sink to drain.
Catalog No. 50-334-54 Supplier Electron Microscopy Sciences Supplier No. 6414301
RDO Decalcifier was developed with the basic premise of making your work routine quicker and easier without sacraficing the quality and integrity of your samples. It is a remarkably effective multipurpose decacifier that can be tailored to suit your specific lab routine. RDO is safe, non-toxic, and non-corrisive. It is dilutable, microwavable, biodegradable, and translucent.
RDO Decalcifier offers crisp nuclear staining and definition, gives distinguishable chromatin and shows great surface decalcification. It is safe enough to be disposed down lab sink to drain.