Learn More
Electron Microscopy Sciences RDO – Gold Decalcifier, 1 gal
SDP
Catalog No. 5033456
Encompass_Preferred
RDO and RDO Gold were developed with the basic premise of making your work routine quicker and easier without sacraficing the quality and integrity of your samples. RDO and RDO Gold are remarkably effective multipurpose decacifiers that can be tailored to suit your specific lab routine.
Catalog No. 50-334-56 Supplier Electron Microscopy Sciences Supplier No. 6414440
RDO and RDO Gold were developed with the basic premise of making your work routine quicker and easier without sacraficing the quality and integrity of your samples. RDO and RDO Gold are remarkably effective multipurpose decacifiers that can be tailored to suit your specific lab routine.