Learn More
Medchemexpress LLC RdRP-IN-2 | 2863657-16-1 | 98.0% | C33H30N2O5S | 10MG
SDP
Catalog No. 5000260518
Encompass_Preferred
RdRP-IN-2 is a small-molecule inhibitor of RNA-dependent RNA polymerase (RdRp) with reported activity against SARS-CoV-2 RdRp (IC50 = 41.2 μM). The compound is provided for research use and is characterized by the molecular formula C33H30N2O5S and a molecular weight of 566.67; available in multiple quantities and formulation options for in vitro studies.
  • Inhibits RNA-dependent RNA polymerase (RdRp) activity.
  • Reported IC50 of 41.2 μM against SARS-CoV-2 RdRp.
  • Available in multiple pack sizes and solution formats for flexible use.
  • High purity (>98% by HPLC) for analytical and biological assays.
  • Suitable for in vitro antiviral and mechanistic research.
  • Follow supplier safety data sheet for storage and handling instructions.
Catalog No. 50-002-60518 Supplier Medchemexpress LLC Supplier No. HY13944210MG
RdRP-IN-2 is a small-molecule inhibitor of RNA-dependent RNA polymerase (RdRp) with reported activity against SARS-CoV-2 RdRp (IC50 = 41.2 μM). The compound is provided for research use and is characterized by the molecular formula C33H30N2O5S and a molecular weight of 566.67; available in multiple quantities and formulation options for in vitro studies.
  • Inhibits RNA-dependent RNA polymerase (RdRp) activity.
  • Reported IC50 of 41.2 μM against SARS-CoV-2 RdRp.
  • Available in multiple pack sizes and solution formats for flexible use.
  • High purity (>98% by HPLC) for analytical and biological assays.
  • Suitable for in vitro antiviral and mechanistic research.
  • Follow supplier safety data sheet for storage and handling instructions.