Learn More
Medchemexpress LLC Rdrp-in-2 | 2863657-16-1 | 98.01% | 566.67 | 100 MG
SDP
Catalog No. 5000350862
Encompass_Preferred
RdRP-IN-2 is a potent RNA dependent RNA polymerase (RdRp) inhibitor, demonstrating significant activity against SARS-CoV-2 RdRp with an IC50 of 41.2 μM. This compound also exhibits inhibitory effects on Feline coronavirus (FIPV) replication, highlighting its broad potential in antiviral research against RNA viruses.
  • Potent RNA dependent RNA polymerase (RdRp) inhibitor
  • Significantly inhibits SARS-CoV-2 RdRp
  • Exhibits inhibitory effects on Feline coronavirus (FIPV) replication
  • Useful for antiviral research against RNA viruses
Catalog No. 50-003-50862 Supplier Medchemexpress LLC Supplier No. HY139442100MG
RdRP-IN-2 is a potent RNA dependent RNA polymerase (RdRp) inhibitor, demonstrating significant activity against SARS-CoV-2 RdRp with an IC50 of 41.2 μM. This compound also exhibits inhibitory effects on Feline coronavirus (FIPV) replication, highlighting its broad potential in antiviral research against RNA viruses.
  • Potent RNA dependent RNA polymerase (RdRp) inhibitor
  • Significantly inhibits SARS-CoV-2 RdRp
  • Exhibits inhibitory effects on Feline coronavirus (FIPV) replication
  • Useful for antiviral research against RNA viruses