Learn More
AccuStandard, Inc RDX STANDARD MIX 1.0MG/ML
SDP
Catalog No. NC9339497
Encompass
RDX . 1 mL. 1000 ?g/mL in MeOH:AcCN (1:1). 121-82-4
Catalog No. NC9339497 Supplier AccuStandard, Inc Supplier No. M833005
RDX . 1 mL. 1000 ?g/mL in MeOH:AcCN (1:1). 121-82-4