Learn More
MSC Zep Detergent Disinfectant, Ready-to-Use, Citrus Scent, Virucidal, Tuberculocidal, 5 gal
Catalog No. 502867598
Encompass
Zep Spirit II Detergent Disinfectant is a ready-to-use, citrus-scented liquid formulated for general cleaning and disinfection of hard, non-porous surfaces. It is designed to control cross-contamination in a wide range of environments, including healthcare, food service, and residential settings.
  • Disinfectant, deodorizer, cleaner, virucide, tuberculocidal, and fungicide
  • Effective against HIV-1, HBV, Influenza A (H1N1), Norovirus, Rabies Virus, and canine parvovirus
  • Kills Paramyxovirus (Mumps), Rhinovirus type 39, and Rotavirus
  • Powerful detergent system cuts through grease and heavy soil
  • Suitable for use on floors, walls, metal, stainless steel, glazed porcelain, glazed ceramic tile, and plastics
Catalog No. 50-286-7598 Supplier MSC Supplier No. 17644311
Zep Spirit II Detergent Disinfectant is a ready-to-use, citrus-scented liquid formulated for general cleaning and disinfection of hard, non-porous surfaces. It is designed to control cross-contamination in a wide range of environments, including healthcare, food service, and residential settings.
  • Disinfectant, deodorizer, cleaner, virucide, tuberculocidal, and fungicide
  • Effective against HIV-1, HBV, Influenza A (H1N1), Norovirus, Rabies Virus, and canine parvovirus
  • Kills Paramyxovirus (Mumps), Rhinovirus type 39, and Rotavirus
  • Powerful detergent system cuts through grease and heavy soil
  • Suitable for use on floors, walls, metal, stainless steel, glazed porcelain, glazed ceramic tile, and plastics