Learn More
MSC Zep Hardwood and Laminate Floor Cleaner, 32 oz
Catalog No. 502818792
Encompass
Zep Hardwood and Laminate Floor Cleaner is a concentrated formula designed for floor care, effectively removing spots, stains, and scuffs. It restores shine to varnish, acrylic, and polyurethane-finished floors, molding, cabinet doors, and other wood surfaces.
  • Removes spots, stains, and scuffs
  • Restores shine to varnish, acrylic, and polyurethane-finished floors, molding, cabinet doors, and other wood surfaces
  • Safe for use on varnish, acrylic, and polyurethane-finished wood and laminate floors, baseboards, cabinet doors, and crown molding
  • Not for use on tile or floors with grout, oil finished wood floors, or floors that require waxing
  • Ready-to-use formula, no dilution required
  • Dries fast with no rinsing needed
  • Non-toxic and phosphate-free
  • Leaves a fresh scent
  • Has a pH range of 8.5–9.0
Catalog No. 50-281-8792 Supplier MSC Supplier No. 17689977
Zep Hardwood and Laminate Floor Cleaner is a concentrated formula designed for floor care, effectively removing spots, stains, and scuffs. It restores shine to varnish, acrylic, and polyurethane-finished floors, molding, cabinet doors, and other wood surfaces.
  • Removes spots, stains, and scuffs
  • Restores shine to varnish, acrylic, and polyurethane-finished floors, molding, cabinet doors, and other wood surfaces
  • Safe for use on varnish, acrylic, and polyurethane-finished wood and laminate floors, baseboards, cabinet doors, and crown molding
  • Not for use on tile or floors with grout, oil finished wood floors, or floors that require waxing
  • Ready-to-use formula, no dilution required
  • Dries fast with no rinsing needed
  • Non-toxic and phosphate-free
  • Leaves a fresh scent
  • Has a pH range of 8.5–9.0