Learn More
Strem, An Ascensus Company CAS# 14220-21-4. 250mg. Rhenium pentacarbonyl bromide, 98%. MFCD00013295
SDP
Catalog No. 502045385
Encompass
CAS# 14220-21-4. 250mg. Rhenium pentacarbonyl bromide, 98%. MFCD00013295. Molecular Weight: 406.16. Molecular Formula: Re(CO)5Br. Color/form: off-white xtl. Strem# 75-0150. http://www.strem.com/catalog/v/75-0150/
Catalog No. 50-204-5385 Supplier Strem, An Ascensus Company Supplier No. 750150250MG
CAS# 14220-21-4. 250mg. Rhenium pentacarbonyl bromide, 98%. MFCD00013295. Molecular Weight: 406.16. Molecular Formula: Re(CO)5Br. Color/form: off-white xtl. Strem# 75-0150. http://www.strem.com/catalog/v/75-0150/