Learn More
Mopec Re-Print Tabs
SDP
Catalog No. 22446320 Shop All Mopec Products
Change view
Click to view available options
Quantity:
500/Roll
Catalog No. Quantity
22-446-320 500/Roll
1 options
Catalog No. 22-446-320 Supplier Mopec Supplier No. FS465
Add to Cart
Add to Cart
Adhesive-backed tabs cover and correct poor or unwanted fingerprint impressions on record cards, allowing a new print to be placed over the affected area. They work with various fingerprint inking systems and are supplied in dispenser-style rolls for convenient access during processing.
Construction
  • Adhesive-backed
  • Packaged in a hassle-free dispenser box
Quantity
  • 500 per roll
Specifications
Product Type Re-Print Tab
Quantity 500/Roll