Learn More
Bioworld Re-Probe Solution western Blots, 125 mL
Catalog No. 50198553
Encompass_Preferred
Re-Probe Solution, Western BlotsRe-Probe is supplied as 5X. The 125ml kit sufficient for 25 blots. Not recommended for chromogenic substrates.
Catalog No. 50-198-553 Supplier Bioworld Supplier No. 209600081
Re-Probe Solution, Western BlotsRe-Probe is supplied as 5X. The 125ml kit sufficient for 25 blots. Not recommended for chromogenic substrates.