Learn More
Pluriselect Re-Strainer (In-Line-Strainer) 300 µm - Sterile
Catalog No. NC2877598
Encompass_Preferred
The Re-Strainer is a filtration device for high volumes of liquid to exclude or concentrate particles from liquids with the possibility of particle recovery. The 6 ml recovery reservoir can be accessed by unscrewing the cap. The Re-Strainer has two female luer-lock-ports.These ports can be used to connect several Re-Strainer to a filter cascade (male to male luer-lock connector) or integrate the Re-Strainer as an in-line-strainer via luer-lock tube adaptor.The mesh is an integrated part of the housing to give the device maximum mechanical stability.
Catalog No. NC2877598 Supplier Pluriselect Supplier No. 437530040
The Re-Strainer is a filtration device for high volumes of liquid to exclude or concentrate particles from liquids with the possibility of particle recovery. The 6 ml recovery reservoir can be accessed by unscrewing the cap. The Re-Strainer has two female luer-lock-ports.These ports can be used to connect several Re-Strainer to a filter cascade (male to male luer-lock connector) or integrate the Re-Strainer as an in-line-strainer via luer-lock tube adaptor.The mesh is an integrated part of the housing to give the device maximum mechanical stability.