Learn More
Pluriselect Re-Strainer (In-Line-Strainer) 1 um mesh-size, 20 pcs. sterile
Catalog No. NC2020977
Encompass_Preferred
The Re-Strainer (In-Line-Strainer) has a 6 ml recovery reservoir which can be accessed by unscrewing the cap. The Re-Strainer has two female luer-lock-ports. These ports can be used to connect several Re-Strainer to a filter cascade (male to male luer-lock connector) or integrate the Re-Strainer as an in-line-strainer via luer-lock tube adaptor. You can choose from 15 different mesh sizes (1 um to 500 um)
Catalog No. NC2020977 Supplier Pluriselect Supplier No. 437500140
The Re-Strainer (In-Line-Strainer) has a 6 ml recovery reservoir which can be accessed by unscrewing the cap. The Re-Strainer has two female luer-lock-ports. These ports can be used to connect several Re-Strainer to a filter cascade (male to male luer-lock connector) or integrate the Re-Strainer as an in-line-strainer via luer-lock tube adaptor. You can choose from 15 different mesh sizes (1 um to 500 um)