Learn More
Yamato Scientific America, Inc. Re202 Rotary Evaporator Set With Bo302 Oil Bath And Vertical Condenser B 115V
Catalog No. NC1769320 Shop All Yamato Scientific America Inc. Products
Encompass_Preferred
Standard rotary evaporator with slide jack mechanism for convenient height adjustment. Set includes BO302-A oil bath (115V) and glassware set B with vertical condenser. Market leader in rotation speed range at 5~315 rpm. Designed with set inversion function where glassware and bath can be set on either side, left or right, depending on user’s dominant hand and installation location. Standard equipped with condenser insulation kit to suppress condensation on the condenser’s surface. Available in 220V and in combination with water bath.
Catalog No. NC1769320 Supplier Yamato Scientific America, Inc. Supplier No. RE202BOA
Standard rotary evaporator with slide jack mechanism for convenient height adjustment. Set includes BO302-A oil bath (115V) and glassware set B with vertical condenser. Market leader in rotation speed range at 5~315 rpm. Designed with set inversion function where glassware and bath can be set on either side, left or right, depending on user’s dominant hand and installation location. Standard equipped with condenser insulation kit to suppress condensation on the condenser’s surface. Available in 220V and in combination with water bath.