Learn More
Genesee Scientific Corporation Genesee Scientific Reach Tip 1000µl, Ergonomic Design, RNase/DNase Free, 1000 µl
SDP
Catalog No. NC1073831
Encompass
Genesee Scientific Reach Tips are designed for precise liquid handling in scientific applications. Engineered with an ergonomic design and extended length, they prevent pipettor shaft contamination and enhance user comfort, reducing the risk of Repetitive Strain Injury (RSI). Made from 100% virgin polypropylene, these low-retention tips maximize sample recovery while being certified free of contaminants. Their universal fit ensures compatibility with most pipettors, making them an ideal choice for laboratories.
Catalog No. NC1073831 Supplier Genesee Scientific Corporation Supplier No. 24165RL
Genesee Scientific Reach Tips are designed for precise liquid handling in scientific applications. Engineered with an ergonomic design and extended length, they prevent pipettor shaft contamination and enhance user comfort, reducing the risk of Repetitive Strain Injury (RSI). Made from 100% virgin polypropylene, these low-retention tips maximize sample recovery while being certified free of contaminants. Their universal fit ensures compatibility with most pipettors, making them an ideal choice for laboratories.