Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC ReACp53 scrambled peptide TFA | 63.7% | 2617.13 | 5 MG
SDP

Catalog No. 5000426381
Encompass_Preferred

ReACp53 scrambled peptide TFA serves as a negative control for ReACp53 (HY-P0121) and is utilized in the study of ovarian cancer.

  • Functions as a negative control for ReACp53
  • Molecular formula: C108H206N52O24.XC2HF3O2
  • Sequence: Arg-Arg-Arg-Arg-Arg-Arg-Arg-Arg-Arg-Arg-Ile-Thr-Thr-Arg-Pro-Ile-Leu-Leu-Glu (TFA salt)
  • Shortened sequence: RRRRRRRRRRITTRPILLE
  • Targets MDM-2/p53
  • Pathway involved: Apoptosis
  • Peptide content: 63.69%

Catalog No. 50-004-26381 Supplier Medchemexpress LLC Supplier No. HYP0121B5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ReACp53 scrambled peptide TFA serves as a negative control for ReACp53 (HY-P0121) and is utilized in the study of ovarian cancer.

  • Functions as a negative control for ReACp53
  • Molecular formula: C108H206N52O24.XC2HF3O2
  • Sequence: Arg-Arg-Arg-Arg-Arg-Arg-Arg-Arg-Arg-Arg-Ile-Thr-Thr-Arg-Pro-Ile-Leu-Leu-Glu (TFA salt)
  • Shortened sequence: RRRRRRRRRRITTRPILLE
  • Targets MDM-2/p53
  • Pathway involved: Apoptosis
  • Peptide content: 63.69%

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.