Learn More
Chemglass Life Sciences Reaction Block, 4 Dram (16mL) Vial, 21mm x 70mm, Red, 21 Position, 32mm Hole Depth
SDP
Catalog No. CG199106
Encompass_Preferred
Reaction Block, 4 Dram (16mL) Vial, 21mm x 70mm, Red, 21 Position, 32mm Hole Depth.Excellent heat transferWon't interfere with magnetic stirringHoles spaced for optimum stirring of all vialsColor-anodized for durability and easy identificationRound reaction blocks are designed to fit directly on 135mm diameter, round top hot plate stirrers. Aluminum construction offers excellent heat transfer without interfering with magnetic stirring. All blocks are anodized for increased strength, durability and chemical resistance. A thermowell is provided for insertion of a temperature sensor or contact thermometer. Available for scintillation vials and dram vials.
Catalog No. CG199106 Supplier Chemglass Life Sciences Supplier No. CG199106
Reaction Block, 4 Dram (16mL) Vial, 21mm x 70mm, Red, 21 Position, 32mm Hole Depth.Excellent heat transferWon't interfere with magnetic stirringHoles spaced for optimum stirring of all vialsColor-anodized for durability and easy identificationRound reaction blocks are designed to fit directly on 135mm diameter, round top hot plate stirrers. Aluminum construction offers excellent heat transfer without interfering with magnetic stirring. All blocks are anodized for increased strength, durability and chemical resistance. A thermowell is provided for insertion of a temperature sensor or contact thermometer. Available for scintillation vials and dram vials.