Learn More
Chemglass Life Sciences OptiBlock Parallel Synthesis Reaction Block Start Up Kit
SDP
Catalog No. 507513683
Encompass_Preferred
OptiBlock Parallel Synthesis Reaction Block Start Up Kit. • Seals Multiple Vessels Effectively • Wide Range of Vessel Sizes • Provides Syringe Access to all Vessels • Works at Boiling Point • Glove Box and Automation Friendly • Uses Existing Hot Plate Stirrers • Works with Aggressive SolventsThe OPTIBLOCK is a simple, easy to use tool for medicinal and process chemists. The block accepts up to eight different vial sizes and fits directly onto a hot plate stirrer having a 135mm top plate diameter.PLEASE NOTE: The OP-5000-K1 Start Up Kit Consists of 1 each of the following: OP-5000-01, OP-5000-10, OP-5000-12, OP-5000-16, OP-6600-05, OP-6600-07 and OP-6600-11 and 20 each of OP-5000-60.
Catalog No. 50-751-3683 Supplier Chemglass Life Sciences Supplier No. OP5000K1
OptiBlock Parallel Synthesis Reaction Block Start Up Kit. • Seals Multiple Vessels Effectively • Wide Range of Vessel Sizes • Provides Syringe Access to all Vessels • Works at Boiling Point • Glove Box and Automation Friendly • Uses Existing Hot Plate Stirrers • Works with Aggressive SolventsThe OPTIBLOCK is a simple, easy to use tool for medicinal and process chemists. The block accepts up to eight different vial sizes and fits directly onto a hot plate stirrer having a 135mm top plate diameter.PLEASE NOTE: The OP-5000-K1 Start Up Kit Consists of 1 each of the following: OP-5000-01, OP-5000-10, OP-5000-12, OP-5000-16, OP-6600-05, OP-6600-07 and OP-6600-11 and 20 each of OP-5000-60.