Learn More
Chemglass Life Sciences Reaction Block Vial Holder, 96-Well, with 8x30mm Vials, Complete
SDP
Catalog No. 501539695
Encompass_Preferred
Our reaction block vial holder has the same footprint as standard 96-well plates and is compatible with 96-well automated work stations. The holder is made of polypropylene, holds 8x30mm OD borosilicate vials, and is reusable. Standard 8x12 alpha-numeric format: A-H, 1-12. Accepts standard SBS (7mm round diameter plug) cap mat; made with 100% premium silicone with PTFE spray coating, available in packs of 5, non- or pre-slit. An ideal solution for: High throughput screening, solubility screening, polymorph screening, powder dispensing, reaction screening, or biological Testing.Compatible with: Unchained Labs (Freeslate), CM2 and CM3 Chemspeed, Waters Acquity UPLC, Shimadzu Prominence, Agilent BenchCel.
Catalog No. 50-153-9695 Supplier Chemglass Life Sciences Supplier No. CG191401
Our reaction block vial holder has the same footprint as standard 96-well plates and is compatible with 96-well automated work stations. The holder is made of polypropylene, holds 8x30mm OD borosilicate vials, and is reusable. Standard 8x12 alpha-numeric format: A-H, 1-12. Accepts standard SBS (7mm round diameter plug) cap mat; made with 100% premium silicone with PTFE spray coating, available in packs of 5, non- or pre-slit. An ideal solution for: High throughput screening, solubility screening, polymorph screening, powder dispensing, reaction screening, or biological Testing.Compatible with: Unchained Labs (Freeslate), CM2 and CM3 Chemspeed, Waters Acquity UPLC, Shimadzu Prominence, Agilent BenchCel.